Merck
  • Multiplex ligation-dependent probe amplification and array comparative genomic hybridization analyses for prenatal diagnosis of cytogenomic abnormalities.
Multiplex ligation-dependent probe amplification and array comparative genomic hybridization analyses for prenatal diagnosis of cytogenomic abnormalities.
Molecular cytogenetics (2014-12-23)
Zhiyong Xu, Qian Geng, Fuwei Luo, Fang Xu, Peining Li, Jiansheng Xie
ABSTRACT
The aims of this study were to evaluate the clinical utility of multiplex ligation-dependent probe amplification (MLPA) and array comparative genomic hybridization (aCGH) analyses on prenatal cases and to review prenatal ultrasound findings of cytogenomic syndromes. Of the 54 prenatal cases analyzed, cytogenomic abnormalities were characterized in 14 cases. In four fetuses with abnormal ultrasound findings, a 40.701 Mb duplication of 8q22.3-q24.3 and a 23.839 Mb deletion of 7q33-q36.3 derived from a paternal balanced translocation, a de novo 13.062 Mb deletion of 11q24.1-q25 for Jacobsen syndrome, a de novo 19.971 Mb deletion of 7q11.23-q21.3 for type 1 split-hand/foot malformation (SHFM1), and a de novo 28.909 Mb duplication of 3q21.1-q25.1 were detected. A 699.8 Kb deletion at 5p15.33 for Cri du Chat syndrome was confirmed in a fetus with abnormal MLPA result. A fetus with abnormal maternal screening was detected with a de novo distal 1.747 Mb duplication at 2q37.1-q37.2 and a 6.664 Mb deletion at 2q37.2-q37.3. Of the eight cases referred by history of spontaneous abortions, derivative chromosomes 11 from paternal carriers of a balanced 8q/11q and a 10q/11q translocation were noted in two cases, simple aneuploids of trisomy 2 and trisomy 21 were seen in three cases, and compound aneuploids of two or three chromosomes were found in three cases. Post-test genetic counseling was performed with detailed genomic information and well characterized postnatal syndromic features. These results demonstrated that coupling MLPA screening and aCGH analysis are a cost-effective approach to detect cytogenomic abnormalities in a prenatal setting. The aCGH analysis provided not only genomic maps of breakpoints and gene content of imbalanced regions but also better inference of related phenotypes for genetic counseling. Prenatal ultrasound findings reported in the literature for Jacobsen syndrome, SHFM and Cri du Chat syndrome were summarized for use as diagnostic references.