Merck
  • Nuclear protein IK undergoes dynamic subcellular translocation and forms unique nuclear bodies during the cell cycle.
Nuclear protein IK undergoes dynamic subcellular translocation and forms unique nuclear bodies during the cell cycle.
Biomarker research (2013-11-21)
Liyan Hu, Feikun Yang, Xianan Liu, Dazhong Xu, Wei Dai
ABSTRACT
IK is a nuclear protein containing a unique domain named RED due to the presence of a repetitive arginine (R), aspartic (E), and glutamic acid (D) sequence. To date, the function of this protein remains largely unknown despite of a couple of previous studies in the literature. Here we report that depletion of IK via RNA interference results in mitotic arrest. We also demonstrate that IK undergoes dynamic translocation during interphase and mitosis. In particular, IK is primarily present in some interphase cells as nuclear foci/bodies which do not co-localize with nucleoli, PMA bodies and Cajal bodies. Pull-down analysis coupled with mass spectrometry reveals that IK is associated with DHX15, a putative ATP-dependent RNA helicase. Our results strongly suggest that IK may participate in pre-mRNA splicing and that it may be a useful biomarker for a new nuclear structure in the cell.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ik