Merck
Association between ABCC2 gene haplotypes and tenofovir-induced proximal tubulopathy.
The Journal of infectious diseases (2006-11-04)
Hassane Izzedine, Jean-Sebastien Hulot, Eric Villard, Catherine Goyenvalle, Stephanie Dominguez, Jade Ghosn, Marc Antoine Valantin, Philippe Lechat, And Gilbert Deray
ABSTRACT
Tenofovir disoproxil fumarate (TDF) may induce renal proximal tubulopathy (rPT). There are no data on pharmacogenomic predictors of rPT in the genes encoding the multidrug-resistance protein (MRP) 2 and MRP4 transporters. Mutational screening of the genes for MRP2 (ABCC2) and MRP4 (ABCC4) was performed using genomic DNA from 13 human immunodeficiency virus type 1 (HIV-1)-infected patients (group 1) presenting with TDF-induced rPT. Concomitantly, 17 unrelated HIV-1-infected patients who had received TDF therapy and who did not have rPT (group 2) were included in a case-control analysis, to assess the influence of single-nucleotide polymorphisms (SNPs) identified in ABCC2 and ABCC4. Six SNPs were identified in ABCC2. A significant allelic association between the 1249 G-->A SNP and TDF-induced rPT was observed (odds ratio, 6.11 [95% confidence interval, 1.19-31.15]; P<.02). ABCC2 haplotypes were significantly associated with the onset of TDF-induced rPT--CATC appeared to be a predisposing haplotype, as it was found in 40.9% of the group 1 case patients and in 13.7% of the group 2 control subjects (P<.01), whereas CGAC appeared to be a protective haplotype, as it was not observed in the group 1 case patients but was present in 20.2% of the group 2 control subjects (P<.01). No association was observed between ABCC4 polymorphism and TDF-induced rPT in the present study. ABCC2 haplotypes are associated with rPT induced by TDF in HIV-1-infected patients.