Merck
  • The nuclear protein PH5P of the inter-alpha-inhibitor superfamily: a missing link between poly(ADP-ribose)polymerase and the inter-alpha-inhibitor family and a novel actor of DNA repair?
The nuclear protein PH5P of the inter-alpha-inhibitor superfamily: a missing link between poly(ADP-ribose)polymerase and the inter-alpha-inhibitor family and a novel actor of DNA repair?
FEBS letters (1999-04-01)
L Jean, J L Risler, T Nagase, C Coulouarn, N Nomura, J P Salier
ABSTRACT
Poly(ADP-ribose)polymerase is a nuclear NAD-dependent enzyme and an essential nick sensor involved in cellular processes where nicking and rejoining of DNA strands are required. The inter-alpha-inhibitor family is comprized of several plasma proteins that all harbor one or more so-called heavy chains designated H1-H4. The latter originate from precursor polypeptides H1P-H4P whose upper two thirds are highly homologous. We now describe a novel protein that includes (i) a so-called BRCT domain found in many proteins involved in DNA repair, (ii) an area that is homologous to the NAD-dependent catalytic domain of poly(ADP-ribose)polymerase, (iii) an area that is homologous to the upper two thirds of precursor polypeptides H1P-H4P and (iv) a proline-rich region with a potential nuclear localization signal. This protein now designated PH5P points to as yet unsuspected links between poly(ADP-ribose)polymerase and the inter-alpha-inhibitor family and is likely to be involved in DNA repair.