Merck
A chemical synthesis of 11-methoxy mitragynine pseudoindoxyl featuring the interrupted Ugi reaction.
Chemical science (2013-07-24)
Jimin Kim, John S Schneekloth, Erik J Sorensen
ABSTRACT
A synthesis of 11-methoxy mitragynine pseudoindoxyl, a new member of the mitragynine class of opioid agonists, from a derivative of the Geissman-Waiss lactone is described. An internal attack of an electron-rich aromatic ring on an electrophilic nitrilium ion and a late-stage construction of the functionalized piperidine ring by the method of reductive cyclization are the pivotal transformations; both ring annulations proceed in a highly diastereoselective fashion. The construction of substituted indoxyl frameworks by the interrupted Ugi method offers an attractive alternative to the strategy of oxidatively rearranging indoles.