- Dihydromevinolin, a potent hypocholesterolemic metabolite produced by Aspergillus terreus.
Dihydromevinolin, a potent hypocholesterolemic metabolite produced by Aspergillus terreus.
The Journal of antibiotics (1981-05-01)
G Albers-Schönberg, H Joshua, M B Lopez, O D Hensens, J P Springer, J Chen, S Ostrove, C H Hoffman, A W Alberts, A A Patchett
PMID7275832
ABSTRACT
A new, potent hypocholesterolemic agent is produced by cultures of Aspergillus terreus. The isolation of the compound and its characterization as 4a,5-dihydromevinolin containing a trans-fused octahydro-naphthalene system are described. Comparative data for dihydromevinolin and mevinolin in three biological assays are given: in vitro inhibition of HMG-CoA reductase, inhibition of sterol synthesis in cell cultures, and inhibition of cholesterol synthesis in vivo in rats.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto