Merck
Effects of drug and chemical pretreatments on biliary excretion of phenylcyclohexene in the rat.
Drug and chemical toxicology (1984-01-01)
F C Law, S Chakrabarti
ABSTRACT
The biliary excretion of 14C-phenylcyclohexene and its metabolites were studied in rats pretreated with an inducer or inhibitor of mixed-function oxidases or with an agent known to deplete liver glutathione. Pretreatment of rats with 3-methylcholanthrene or phenobarbital enhanced the biliary excretion of 14C-phenylcyclohexene and/or its metabolites. Conversely, pretreatment of rats with SKF-525A or piperonyl butoxide decreased the biliary excretion of 14C-phenylcyclohexene and/or its metabolites. In contrast, pretreatment of rats with diethylmaleate did not alter significantly the rate of biliary excretion of radioactivity.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-Phenyl-1-cyclohexene, 95%