Merck
  • Antidepressant-like effect of etazolate, a cyclic nucleotide phosphodiesterase 4 inhibitor--an approach using rodent behavioral antidepressant tests battery.
Antidepressant-like effect of etazolate, a cyclic nucleotide phosphodiesterase 4 inhibitor--an approach using rodent behavioral antidepressant tests battery.
European journal of pharmacology (2012-06-16)
Ankur Jindal, Radhakrishnan Mahesh, Baldev Gautam, Shvetank Bhatt, Dilip Pandey
ABSTRACT
Etazolate, a pyrazolopyridine class derivative is selective inhibitor of type 4 phosphodiesterase (PDE4), an enzyme catalyzes the hydrolysis of cyclic nucleotide viz. cAMP & regulates cAMP signal transduction. Enhancing cAMP signal transduction by inhibition of PDE4 is known to be beneficial in depression disorders. Thus, the present study was designed to investigate thoroughly the antidepressant potential of etazolate using rodent behavioral models of depression. Acute treatment of etazolate (0.25-1 mg/kg, i.p.) exhibited antidepressant-like effects in forced swim test (FST) & tail suspension test (TST) in mice without influencing the baseline locomotion in actophotometer test. Interaction studies of etazolate sub-effective dose (0.12 mg/kg, i.p.), were carried out with sub-effective dose of conventional antidepressants like fluoxetine (5mg/kg, i.p.), venlafaxine (4 mg/kg, i.p.) & desipramine (5 mg/kg, i.p.) in FST. Etazolate at sub-effective dose produced synergistic antidepressant-like effect with conventional antidepressants in the mouse FST. In addition, combined treatment of etazolate & conventional antidepressants had no significant effect on baseline locomotion. Moreover, etazolate (0.5 and 1 mg/kg, i.p.) increased head twitch scores in mice & antagonized the reserpine-induced hypothermia in rats. Chronic treatment (14 days) with etazolate (0.5 and 1 mg/kg, p.o.) & fluoxetine (10 mg/kg, p.o.) significantly reversed the behavioral anomalies induced by bilateral olfactory bulbectomy in rats in modified open field exploration. In conclusion, taken together, our results suggested that etazolate exhibited antidepressant-like activity in acute & chronic rodent models of depression & deserves as a therapeutic tool that could help the conventional pharmacotherapy of depression.