Merck
Variability of microsomal oxidation and porphyrin metabolism in rats.
Bulletin of experimental biology and medicine (2011-01-18)
V E Tseilikman, V E Ryabinin, A S Popova, L I Krupitskaya, A I Sinitsky
ABSTRACT
An inverse relationship between erythropoiesis intensity and microsomal oxidation level has been detected during the early postnatal period in rats with high resistance to hypoxia.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Supelco
Hexobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®