Merck
Effects of beta-cyclodextrins on the enzymatical hydrolysis of chiral dichlorprop methyl ester.
Journal of environmental sciences (China) (2005-11-22)
Yue-Zhong Wen, Shan-shan Zhou, Zhao-hua Fang, Wei-ping Liu
ABSTRACT
The effect of beta-cyclodextrins (beta-CDs) on the enzymatical hydrolysis of chiral dichlorprop methyl ester (DCPPM) was studied. Four kinds of beta-cyclodextrins (beta-cyclodextrin, Partly methylated-CD (PM-beta-CD), hydroxypropyl-cyclodextrin (HP-beta-CD) and carboxymethyl-cyclodextrin (CM-beta-CD)) were used. Compared with 100% DCPPM in the absence of betacyclodextrins, the activity of lipase decreased with the increase of Pbetacyclodextrin and PM-Pbetacyclodextrin. However, CM-beta-clodextrin stimulated the lipase activity. The inhibition effect of Pbeta cyclodextrin and PM-5betacyclodextrin on the hydrolysis of DCPPM is affected by many factors other than degree of the methylation blocking the active site of lipase. UV-Vis and Fourier transform infrared (FTIR) spectroscopy studies of the complexation of aqueous DCPPM with beta-CDs provide fresh insight into the molecular structure of the complex and explain the effects of beta-CDs on enzymatical hydrolysis of chiral DCPPM. Data showed that inclusion complexes had formed by complexation of the CM-beta-CD with DCPPM and the solubility of DCPPM was increased in water, which lead to the increased lipase activity.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Supelco
Dichlorprop-methyl ester, PESTANAL®, analytical standard