Merck
  • Cytotoxicity activity of L-proline analogues of anthraquinone-2-carboxylic acid in breast cancer MCF-7 cells.
Cytotoxicity activity of L-proline analogues of anthraquinone-2-carboxylic acid in breast cancer MCF-7 cells.
Folia histochemica et cytobiologica (2002-02-01)
A Bielawska, K Bielawski, J Pałka
ABSTRACT
Although prolidase [E.C.3.4.13.9] is found in normal cells, substantially increased levels are found in some neoplastic tissues. Prolidase evokes the ability to hydrolyse the imido-bond of various low molecular weight compounds coupled to L-proline. The synthesis of three proline analogues of anthraquinone-2-carboxylic acid (1-3) has been performed. Treatment of these prodrugs with prolidase generated L-proline and the free drug, demonstrating their substrate susceptibility prolidase. The concentrations of 1, 2 and 3 needed to inhibit [1H]thymidine incorporation into DNA by 50% (IC50) in breast cancer MCF-7 cells were found to be 185 +/- 5 microM, 107 +/- 6 microM and 87 +/- 6 microM, respectively, suggesting a lower cytotoxic potency of these compounds compared to Hoechst 33228 (IC50 = 55 +/- 6 microM).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anthraquinone-2-carboxylic acid, 98%