Merck
Recovery and refining of Au by gold-cyanide ion biosorption using animal fibrous proteins.
Applied biochemistry and biotechnology (1998-06-17)
S Ishikawa, K Suyama
ABSTRACT
Animal fibrous proteins (AFPs) such as egg-shell membrane (ESM), chicken feather (CF), wool, silk, or elastin are an intricate network of stable and water-insoluble fibers with high surface area and are abundant bioresources. Every AFP tested was found to accumulate gold-cyanide ion from aqueous solutions in high yield, depending on pH and some other parameters. Gold-cyanide ion is adsorbed by AFP at low pH range, with maximum binding observed at approx pH 2.0. Under the certain conditions, gold-cyanide ion was accumulated up to 8.6, 7.1, 9.8, 2.4, and 3.9% of dry weight on ESM, CF, wool, silk, and elastin, respectively. In the case of ESM, it was found that ESM removed gold-cyanide ion almost quantitatively and almost all the gold uptake by ESM was easily desorbed with 0.1 M NaOH. ESM can be used repeatedly for the process of gold adsorption-desorption. The gold-biosorptive capacity of ESM that was chemically modified with glutaraldehyde was higher than that of control. In column procedure, ESM packed on column removed gold-cyanide ion from the dilute aqueous solution to extremely low concentrations (nondetectable concentration of below 1 ppb).
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Potassium dicyanoaurate(I), 98%
Sigma-Aldrich
Potassium dicyanoaurate(I), 99.95% trace metals basis