Merck
Comparative metabolism and elimination of acetanilide compounds by rat.
Xenobiotica; the fate of foreign compounds in biological systems (1994-10-01)
K L Davison, G L Larsen, V J Feil
ABSTRACT
1. 14C-labelled propachlor, alachlor, butachlor, metolachlor, methoxypropachlor and some of their mercapturic acid pathway metabolites (MAP) were given to rat either by gavage or by perfusion into a renal artery. MAP metabolites were isolated from bile and urine. 2. Rat gavaged with propachlor and methoxypropachlor eliminated 14C mostly in urine, whereas rat gavaged with alachlor, butachlor and metolachlor eliminated 14C about equally divided between urine and faeces. When bile ducts were cannulated, the gavaged rat eliminated most of the 14C in bile for all compounds. The amount of 14C in bile from the propachlor-gavaged rat was less than that for the other acetanilides, with the difference being in the urine. 3. The mercapturic acid metabolites 2-methylsulphinyl-N-(1-methylhydroxyethyl)-N-phenylacetam ide and 2-methylsulphinyl-N-(1-methylmethoxyethyl)-N-phenylacetam ide were isolated from the urine and bile of the methoxypropachlor-gavaged rat. 4. Bile was the major route for 14C elimination when MAP metabolites of alachlor, butachlor and metolachlor were perfused into a renal artery. Urine was the major route for 14C elimination when MAP metabolites of propachlor and methoxypropachlor were perfused. Mercapturic acid conjugates were major metabolites in bile and urine when MAP metabolites were perfused. 5. We conclude that alkyl groups on the phenyl portion of the acetanilide causes biliary elimination to be favoured over urinary elimination.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Supelco
Propachlor, PESTANAL®, analytical standard