Merck
  • Delivery of sesamol-loaded solid lipid nanoparticles to the brain for menopause-related emotional and cognitive central nervous system derangements.
Delivery of sesamol-loaded solid lipid nanoparticles to the brain for menopause-related emotional and cognitive central nervous system derangements.
Rejuvenation research (2011-10-08)
Vandita Kakkar, Anil Kumar Mishra, Krishna Chuttani, Kanwaljit Chopra, Indu Pal Kaur
ABSTRACT
The physicochemical nature of sesamol (logP 1.29; solubility 38.8 mg/mL) substantially enhances its tissue distribution, minimizing its brain delivery. Sesamol-loaded solid lipid nanoparticles (S-SLNs) with an average particle size of 122 nm and an entrapment efficiency of 75.9 ± 2.91% were developed. Biochemical and behavioral paradigms clearly established the superiority of orally administered S-SLNs. The same was confirmed evidently by scintigraphic images of rabbits administered radiolabeled SLNs and confocal microscopy of brain sections of rats administered similarly prepared SLNs with a fluorescent marker. This study indicates the special importance of using phosphatidylcholine (as co-surfactant) in the preparation of SLNs for improving memory deficits. The aim of the present work was to develop sesamol as a therapeutic agent for central nervous system derangements.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sesamol, 98%