Merck
Construction of expression-ready cDNA clones for KIAA genes: manual curation of 330 KIAA cDNA clones.
DNA research : an international journal for rapid publication of reports on genes and genomes (2002-08-10)
Daisuke Nakajima, Noriko Okazaki, Hisashi Yamakawa, Reiko Kikuno, Osamu Ohara, Takahiro Nagase
ABSTRACT
We have accumulated information on protein-coding sequences of uncharacterized human genes, which are known as KIAA genes, through cDNA sequencing. For comprehensive functional analysis of the KIAA genes, it is necessary to prepare a set of cDNA clones which direct the synthesis of functional KIAA gene products. However, since the KIAA cDNAs were derived from long mRNAs (> 4 kb), it was not expected that all of them were full-length. Thus, as the first step toward preparing these clones, we evaluated the integrity of protein-coding sequences of KIAA cDNA clones through comparison with homologous protein entries in the public database. As a result, 1141 KIAA cDNAs had at least one homologous entry in the database, and 619 of them (54%) were found to be truncated at the 5' and/or 3' ends. In this study, 290 KIAA cDNA clones were tailored to be full-length or have considerably longer sequences than the original clones by isolating additional cDNA clones and/or connected parts of additional cDNAs or PCR products of the missing portion to the original cDNA clone. Consequently, 265, 8, and 17 predicted CDSs of KIAA cDNA clones were increased in the amino-, carboxy-, and both terminal sequences, respectively. In addition, 40 cDNA clones were modified to remove spurious interruption of protein-coding sequences. The total length of the resultant extensions at amino- and carboxy-terminals of KIAA gene products reached 97,000 and 7,216 amino acid residues, respectively, and various protein domains were found in these extended portions.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acyl-coenzyme A Synthetase from Pseudomonas sp., ≥2 units/mg protein