Merck
  • Cardiac Fibroblasts Promote Ferroptosis in Atrial Fibrillation by Secreting Exo-miR-23a-3p Targeting SLC7A11.
Cardiac Fibroblasts Promote Ferroptosis in Atrial Fibrillation by Secreting Exo-miR-23a-3p Targeting SLC7A11.
Oxidative medicine and cellular longevity (2022-06-10)
Dishiwen Liu, Mei Yang, Yajun Yao, Shanqing He, Youcheng Wang, Zhen Cao, Huiyu Chen, Yuntao Fu, Huafen Liu, Qingyan Zhao
ABSTRACT
The exact mechanism of atrial fibrillation (AF) has been not well elucidated. Ferroptosis is an iron-dependent cell death due to excessive accumulation of peroxidized polyunsaturated fatty acids. However, the molecular mechanism underlying AF and ferroptosis has never been reported. Here, we established the rapid pacing model in vivo and vitro to investigate the relationship between AF and ferroptosis. In canine model of rapid atrial pacing, the content of malondialdehyde and total ions in the atrial tissue of the Pacing group was significantly increased and the exosome inhibitor GW4869 reduced ferroptosis, fibrosis, and inflammation and improved histological and electrophysiological remodeling. In rapid pacing h9c2 cells, the expression of antioxidative stress genes associated with ferroptosis presented sequential changes and proteins involved in ferroptosis such as FTH1, SLC7A11, and GPX4 were gradually depleted. Furthermore, pacing cardiac fibroblast-derived exosomes (CF-exos) exacerbated ferroptosis in h9c2 cells and pretreated pacing-CF-exos with GW4869 alleviated injury to h9c2 cells. In mechanism, our results demonstrated that pacing-CF-exos highly expressed miR-23a-3p by informatics analysis and experimental verification. Inhibitor-miR-23a-3p protected h9c2 cells from ferroptosis accompanying with upregulation of SLC7A11. In addition, SLC7A11 was shown to be the target gene of miR-23a-3p. In conclusion, our results suggest that CF-exos-miR-23a-3p may promote ferroptosis. The development of AF in a persistent direction could be prevented by intervening with exosomal miRNAs to reduce oxidative stress injury and ferroptosis.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-TSG101 antibody produced in rabbit, IgG fraction of antiserum