Merck
  • The role of MITF and Mcl-1 proteins in the antiproliferative and proapoptotic effect of ciprofloxacin in amelanotic melanoma cells: In silico and in vitro study.
The role of MITF and Mcl-1 proteins in the antiproliferative and proapoptotic effect of ciprofloxacin in amelanotic melanoma cells: In silico and in vitro study.
Toxicology in vitro : an international journal published in association with BIBRA (2020-05-22)
Artur Beberok, Jakub Rok, Zuzanna Rzepka, Krzysztof Marciniec, Stanisław Boryczka, Dorota Wrześniok
ABSTRACT
Mcl-1 is a potent antiapoptotic protein which is amplified in many human cancer, while microphthalmia associated transcription factor (MITF) promotes cell proliferation and has pro-survival role. The study was designed to examine whether the interaction between ciprofloxacin, one of the fluoroquinolones derivative, and MITF/Mcl-1 proteins affects C32 melanoma cells viability, proliferation and induces apoptosis. Preliminary molecular docking studies, Western blot analysis and fluorescence image cytometry were applied to demonstrate the signaling pathway underlying antiproliferative and proapoptotic effect of the drug. In silico analysis showed that ciprofloxacin possess the ability to form complexes with MITF and Mcl-1proteins. This phenomenon was confirmed by in vitro experimental model where the drug was found to decrease MITF and increase Mcl-1 expression at the protein level. Moreover, we found that ciprofloxacin decreases the cell viability and exerts anti-proliferative effect on amelanotic C32 melanoma cells. Image cytometric studies showed that the tested drug induced GSH depletion and apoptosis via intrinsic death pathway leading to DNA fragmentation. Analysis of the cell cycle distribution revealed that ciprofloxacin caused a block in the G2/M phase. This is the first study that characterized the role of MITF and Mcl-1 proteins in the antiproliferative and pro-apoptotic effect of ciprofloxacin towards amelanotic melanoma cells, opening the possibility to use of this drug as a potential agent for the treatment of melanoma.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
L-(+)-2-Amino-3-phosphonopropionic acid