Merck
  • Modulation by angiotensin III of nociception-related and arterial pressure-related neuronal responsiveness in the nucleus reticularis gigantocellularis of the rat.
Modulation by angiotensin III of nociception-related and arterial pressure-related neuronal responsiveness in the nucleus reticularis gigantocellularis of the rat.
Regulatory peptides (1994-03-17)
J Y Chan, H F Tsai, T B Kuo, S H Chan
ABSTRACT
We evaluated possible modulation by angiotensin III (AIII) of the interactive effect of noxious stimuli and elevation in systemic arterial pressure on the responsiveness of neurons in the nucleus reticularis gigantocellularis (NRGC) of the medulla oblongata. Combined extracellular single-neuron recording and microiontophoresis were carried out on male, adult Sprague-Dawley rats anesthetized with pentobarbital sodium. The responsiveness of NRGC neurons to nociception (tail clamp) and/or transient hypertension elicited by phenylephrine (5 micrograms/kg, i.v.), in the absence or presence of AIII, was used as the experimental index. Microiontophoretic application of the heptapeptide suppressed the responses of spontaneously active NRGC neurons to individually delivered nociception or hypertension. Interestingly, the preferential reduction in responsiveness to tail clamp upon simultaneous elevation in arterial pressure was reversed to one that favored nociception in the presence of AIII. These actions of the heptapeptide appeared to be receptor-specific, since they were discernibly blocked by its selective antagonist, Ile7-angiotensin III. Our results reveal that neuropeptides such as AIII may differentially modulate neuronal responsiveness according to the prevailing physiologic input(s) to the central nervous system of the animal.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
[Ile7]-Angiotensin III acetate salt hydrate, ≥95% (HPLC)