Merck
Effects of labor on placental fatty acid β oxidation.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians (2012-08-30)
Hector Mendez-Figueroa, Edward K Chien, Huiling Ji, Nicole L Nesbitt, Sivakama S Bharathi, Eric Goetzman
ABSTRACT
To measure the effect labor exerts on fatty acid (FA) oxidation in term human placentas, and to compare enzymes expression and activity between placenta and liver. Placental samples were collected: (a) scheduled non-labored cesarean section and (b) normal vaginal delivery at or beyond 37 weeks. Long and medium-chain FA oxidation were measured using (3)H-labeled FA, ATP concentration was measured via commercial kit. Activity and expression levels of 11 FA enzymes were measured and results compared to both human and mouse liver. Placentas undergoing labor had significantly decreased palmitate oxidation and ATP levels. Octanoic acid oxidation was 10-fold higher than palmitic acid oxidation. No difference in expression or activity level was detected between the groups. Term human placentas express all the enzymes required to oxidize FA, at a rate 20-fold lower than liver. FA Oxidation is not likely an important placental energy source during labor. Further work is needed to determine the functionality of this pathway in placenta.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Palmitoyl coenzyme A lithium salt, ≥90%
Sigma-Aldrich
Butyryl coenzyme A lithium salt hydrate, ≥90%
Sigma-Aldrich
Octanoyl coenzyme A lithium salt hydrate, ≥95% (HPLC)