Merck
  • Differential antagonism of endomorphin-1 and endomorphin-2 spinal antinociception by naloxonazine and 3-methoxynaltrexone.
Differential antagonism of endomorphin-1 and endomorphin-2 spinal antinociception by naloxonazine and 3-methoxynaltrexone.
Brain research (2000-10-18)
S Sakurada, T Hayashi, M Yuhki, T Fujimura, K Murayama, A Yonezawa, C Sakurada, M Takeshita, J E Zadina, A J Kastin, T Sakurada
ABSTRACT
To determine the role of spinal mu-opioid receptor subtypes in antinociception induced by intrathecal (i.t.) injection of endomorphin-1 and -2, we assessed the effects of beta-funaltrexamine (a selective mu-opioid receptor antagonist) naloxonazine (a selective antagonist at the mu(1)-opioid receptor) and a novel receptor antagonist (3-methoxynaltrexone) using the paw-withdrawal test. Antinociception of i.t. endomorphins and [D-Ala(2), MePhe(4), Gly(ol)(5)]enkephalin (DAMGO) was completely reversed by pretreatment with beta-funaltrexamine (40 mg/kg s.c.). Pretreatment with a variety of doses of i.t. or s.c. naloxonazine 24 h before testing antagonized the antinociception of endomorphin-1, -2 and DAMGO. Judging from the ID(50) values of naloxonazine, the antinociceptive effect of endomorphin-2 was more sensitive to naloxonazine than that of endomorphin-1 or DAMGO. The selective morphine-6beta-glucuronide antagonist, 3-methoxynaltrexone, which blocked endomorphin-2-induced antinociception at each dose (0.25 mg/kg s.c. or 2.5 ng i.t.) that was inactive against DAMGO, did not affect endomorphin-1-induced antinociception but shifted the dose-response curve of endomorphin-2 3-fold to the right. These findings may be interpreted as indicative of the existence of a novel mu-opioid receptor subtype in spinal sites, where antinociception of morphine-6beta-glucuronide and endomorphin-2 are antagonized by 3-methoxynaltrexone. The present results suggest that endomorphin-1 and endomorphin-2 may produce antinociception through different subtypes of mu-opioid receptor.