Merck
  • Electrochemiluminescent aptasensor based on resonance energy transfer system between CdTe quantum dots and cyanine dyes for the sensitive detection of Ochratoxin A.
Electrochemiluminescent aptasensor based on resonance energy transfer system between CdTe quantum dots and cyanine dyes for the sensitive detection of Ochratoxin A.
Talanta (2019-04-07)
Jingwen Gao, Ziyi Chen, Lebao Mao, Wei Zhang, Wei Wen, Xiuhua Zhang, Shengfu Wang
ABSTRACT
In this work, an innovative aptasensor based on electrochemiluminescence resonance energy transfer (ECL-RET) from CdTe quantum dots (QDs) to a cyanine dye (Cy5) fluorophore for the determination of Ochratoxin A (OTA) was fabricated. A strong cathodic ECL emission was obtained by the CdTe QDs modified glassy carbon electrode (GCE). After the immobilization with the capture DNA (cDNA) and the sequential hybridization with the probe DNA-modified Cy5 (pDNA, the aptamer of OTA), the ECL signal enhanced obviously through the ECL-RET. Meanwhile, the spectrum- and distance-related ECL enhancement effect was investigated. When the target OTA was in the presence, the pDNA-Cy5 molecules were released from the electrode surface owing to the specific interaction between OTA and aptamer, resulting in an evident decrease of ECL signal. Under optimal conditions, the developed aptasensor displayed the linear response toward OTA in the wide range of 0.0005-50 ng/mL with a low detection limit of 0.17 pg/mL. With the excellent selectivity, stability and repeatability, the strategy provided an efficient and universal method for the sensitive detection of target in practical application.