Merck
Visual and optical quality outcomes of SMILE and FS-LASIK for myopia in the very early phase after surgery.
BMC ophthalmology (2019-04-10)
Ting Liu, Guanting Lu, Kaijian Chen, Qiuxia Kan, Ji Bai
ABSTRACT
Small incision lenticule extraction (SMILE) and femtosecond laser-assisted in situ keratomileusis (FS-LASIK) are frequently used to treat myopia. However, little is known about the impact on recovery of these approaches in the very early postsurgical phase (within 24 h). To compare the efficacy of these two procedures for the treatment of myopia in the early phase after surgery, differences in visual acuity, OSI (objective scattering index), cutoff for modulation transfer function (MTF), and SR (Strehl ratio) between SMILE and FS-LASIK were evaluated at 0, 2, 4 and 24 h postoperatively using two-way analysis of variance (ANOVA). No significant differences between SMILE and FS-LASIK in the MTF cutoff and SR were found (p > 0.05). However, at 2 h and 4 h after surgery, OSI values in the SMILE group were significantly higher than those in the FS-LASIK group, and visual acuity scores in the SMILE group were significantly poorer than those in the FS-LASIK group (p < 0.05). Regarding subjective symptoms, the number of patients complaining of eye dryness, blurred vision, foreign body sensation and eye soreness in the SMILE group were lower than the number in the FS-LASIK group. In conclusion, visual and optical quality outcomes of FS-LASIK for myopia were better than those of SMILE in the very early phase after surgery, a difference that is attributable to the formation of interface haze. ChiCTR1900021451 .