Merck
  • Effect of New Thiophene-Derived Aminophosphonic Derivatives on Growth of Terrestrial Plants. Part 2. Their Ecotoxicological Impact and Phytotoxicity Test Toward Herbicidal Application in Agriculture.
Effect of New Thiophene-Derived Aminophosphonic Derivatives on Growth of Terrestrial Plants. Part 2. Their Ecotoxicological Impact and Phytotoxicity Test Toward Herbicidal Application in Agriculture.
Molecules (Basel, Switzerland) (2018-12-06)
Diana Rogacz, Jarosław Lewkowski, Zbigniew Malinowski, Agnieszka Matusiak, Marta Morawska, Piotr Rychter
ABSTRACT
Background: The aim of this work was to evaluate phytotoxicity of the thiophene derivatives against three persistent weeds of a high degree of resistance (Galinsogaparviflora Cav.,Rumexacetosa L., and Chenopodiumalbum) as well as their ecotoxicological impact on Heterocypris incongruens. In addition, Aliivibrio fischeri was measured. Two of eight described aminophosphonates, namely dimethyl N-(2-methoxyphenyl)amino(2-thienyl)methylphosphonate (2d) and dimethyl N-(tert-butyl)- (2-thienyl)methylphosphonate (2h), have never been reported before. Methods: The phytotoxicity of tested aminophosphonates toward their potential application as soil-applied herbicides was evaluated according to the OECD 208 Guideline. Ecotoxicological properties of investigated compounds were made using the OSTRACODTOXKITTM and Microtox® tests. Results: Obtained results showed that four aminophosphonates have interesting herbicidal properties and N-(2-methylphenyl)amino- (2-thienyl)methylphosphonate (2a) was found to kill efficiently the most resistant plant Chenopodiumalbum. None of the tested compounds showed important toxicity against Aliivibrio fischeri. However, their toxic impact on Heterocypris incongruens was significantly elevated. Conclusions: The aminophosphonate 2a showed herbicidal potential and it is not toxic against tested bacteria (EC50 over 1000 mg/L). It was found to be moderately toxic against ostracods [mortality 48% at 10 mg/kg of soil dry weight (s.d.w.)] and this problem should be solved by the use of the controlled release from a polymeric carrier.