Informazioni su questo articolo
feature
capacity 0.2 ‑ 2.0 mL, lid, to hold 24 x Adaptors (Models 2023, 2024, 2031)
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1195-A
parameter
31,514 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MIKRO 220/220R
Legal Information
1 of 1
Questo articolo | |||
|---|---|---|---|
| manufacturer/tradename Hettich 1195-A | manufacturer/tradename Hettich 1154-L | manufacturer/tradename Hettich 1016 | manufacturer/tradename Hettich 1641 |
| compatibility for use with centrifuge models MIKRO 220/220R | compatibility for use with centrifuge models MIKRO 220/220R | compatibility for use with centrifuge models MIKRO 220/220R | compatibility for use with 1016, 1137 (rotors) |
| parameter 31,514 max. RCF | parameter 18,516 max. RCF | parameter 4,025 max. RCF | parameter - |
| feature capacity 0.2 ‑ 2.0 mL, to hold 24 x Adaptors (Models 2023, 2024, 2031), lid | feature lid, to hold 24 x 1.5/2.0 mL tubes | feature capacity 7 ‑ 50 mL, to hold 6 x 50 mL tubes | feature Holds 50 mL tubes |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 3 ea |
Still not finding the right product?
Explore all of our products under Hettich® MIKRO 220/220R centrifuge accessories
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?