Informazioni su questo articolo
Formula empirica (notazione di Hill):
C32H28N4O3
Numero CAS:
Peso molecolare:
516.59
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutartiassay
≥98% (HPLC)
form
powder
optical activity
[α]/D +128 to +140 (c = 1, DCM)
color
white to beige
solubility
DMSO: 5 mg/mL (clear solution)
storage temp.
room temp
SMILES string
Cc1cccc(NC(=O)N[C@@H]2N=C(c3ccccc3)c4ccccc4N(CC(=O)c5ccccc5C)C2=O)c1
InChI
1S/C32H28N4O3/c1-21-11-10-15-24(19-21)33-32(39)35-30-31(38)36(20-28(37)25-16-7-6-12-22(25)2)27-18-9-8-17-26(27)29(34-30)23-13-4-3-5-14-23/h3-19,30H,20H2,1-2H3,(H2,33,35,39)/t30-/m0/s1
InChI key
YCXFHPUBGMMWJQ-PMERELPUSA-N
Biochem/physiol Actions
YM022 is a very potent, selective antagonist of the gastrin/cholecystokinin (CCK)-B receptor.
YM022 is a very potent, selective antagonist of the gastrin/cholecystokinin (CCK)-B receptor. The Ki value for CCKB is 68 pM vs 63 nM for CCKA. In rats, YM022 inhibits pentagastrin-induced gastric emptying with an ED50 or 7.8 nM/kg.
Features and Benefits
This compound is featured on the Cholecystokinin and Gastrin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Classe di stoccaggio
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?