Vai a
M2683
Media bottles, wide mouth, Lab 45
capacity 500 mL
Sinonimo/i:
bottle wide mouth, wide mouth bottle, wide mouth bottles
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
A voi/SKUDisponibilitàPrezzo
12 ea
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
271,00 €
Informazioni su questo articolo
NACRES:
NB.17
UNSPSC Code:
41121800
Capacity:
500 mL
Material:
borosilicate glass bottle, polypropylene cap
Sterility:
non-sterile
Feature:
graduations, with cap
Packaging:
pack of 12 ea
271,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
borosilicate glass bottle, polypropylene cap
joint
Joints threaded (45)
description
Autoclavable
sterility
non-sterile
feature
graduations, with cap
packaging
pack of 12 ea
manufacturer/tradename
WHEATON®
capacity
500 mL
General description
Graduated safety coated bottles are ideal for storing fluids, dry chemicals, and other materials in the lab and help protect against broken glass. The strong, slip resistant plastisol coating reduces potential breakage and exposure to lab personnel handling hazardous material. Manufactured from Wheaton glass (meets USP Type I standards). Fluids are protected because the glass has a low alkali content. Wide opening, special lip and sloping shoulders facilitate pouring and cleaning. Includes a white polypropylene screw cap with a wedge-shaped inner sealing ring and polypropylene pour ring.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
Z250201M2808Z263206
material
borosilicate glass bottle, polypropylene cap
material
(PTFE faced LDPE liner), amber soda-lime glass bottle, round bottle, white polypropylene cap, wide-mouth bottle
material
borosilicate glass bottle, polypropylene cap, round bottle
material
round bottle, soda-lime glass bottle, vinyl flat liner, white polypropylene cap
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
feature
graduations, with cap
feature
graduations: no, with cap
feature
graduations, with cap
feature
graduations: no, with cap
capacity
500 mL
capacity
500 mL
capacity
1000 mL
capacity
16 oz (500 mL)
packaging
pack of 12 ea
packaging
case of 12 ea
packaging
pack of 12 ea
packaging
pkg of 24 ea
joint
Joints threaded (45)
joint
Joints threaded neck (53-400)
joint
Joints threaded neck (45)
joint
Joints threaded neck (63-400)
Still not finding the right product?
Explore all of our products under Media bottles, wide mouth, Lab 45
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)