Merck
  • Synthesis and biological activity of hydroxylated derivatives of linoleic acid and conjugated linoleic acids.
Synthesis and biological activity of hydroxylated derivatives of linoleic acid and conjugated linoleic acids.
Chemistry and physics of lipids (2009-01-17)
Zhen Li, Van H Tran, Rujee K Duke, Michelle C H Ng, Depo Yang, Colin C Duke
ABSTRACT
Allylic hydroxylated derivatives of the C18 unsaturated fatty acids were prepared from linoleic acid (LA) and conjugated linoleic acids (CLAs). The reaction of LA methyl ester with selenium dioxide (SeO(2)) gave mono-hydroxylated derivatives, 13-hydroxy-9Z,11E-octadecadienoic acid, 13-hydroxy-9E,11E-octadecadienoic acid, 9-hydroxy-10E,12Z-octadecadienoic acid and 9-hydroxy-10E,12E-octadecadienoic acid methyl esters. In contrast, the reaction of CLA methyl ester with SeO(2) gave di-hydroxylated derivatives as novel products including, erythro-12,13-dihydroxy-10E-octadecenoic acid, erythro-11,12-dihydroxy-9E-octadecenoic acid, erythro-10,11-dihydroxy-12E-octadecenoic acid and erythro-9,10-dihydroxy-11E-octadecenoic acid methyl esters. These products were purified by normal-phase short column vacuum chromatography followed by high-performance liquid chromatography (HPLC). Their chemical structures were characterized by liquid chromatography-mass spectrometry (LC-MS) and nuclear magnetic resonance spectroscopy (NMR). The allylic hydroxylated derivatives of LA and CLA exhibited moderate in vitro cytotoxicity against a panel of human cancer cell lines including chronic myelogenous leukemia K562, myeloma RPMI8226, hepatocellular carcinoma HepG2 and breast adenocarcinoma MCF-7 cells (IC(50) 10-75 microM). The allylic hydroxylated derivatives of LA and CLA also showed toxicity to brine shrimp with LD(50) values in the range of 2.30-13.8 microM. However these compounds showed insignificant toxicity to honeybee at doses up to 100 microg/bee.
Sigma-Aldrich
Selenium dioxide, 99.999% trace metals basis
Sigma-Aldrich
Selenium dioxide, ≥99.9% trace metals basis
Sigma-Aldrich
Selenium dioxide, reagent grade, powder, 98%
Sigma-Aldrich
Selenium dioxide, ReagentPlus®, powder, 99.8% trace metals basis