Skip to Content
Merck
All Photos(1)

Documents

V000180

Sigma-Aldrich

Malachite Green Carbinol base

95.0%

Synonym(s):

Solvent Green 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N2O
CAS Number:
Molecular Weight:
346.47
EC Number:
MDL number:
PubChem Substance ID:

Assay

95.0%

mp

112-114 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)C(O)(c2ccccc2)c3ccc(cc3)N(C)C

InChI

1S/C23H26N2O/c1-24(2)21-14-10-19(11-15-21)23(26,18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,26H,1-4H3

InChI key

LXHOGENDFZKPSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service