Skip to Content
Merck
All Photos(1)

Documents

1154605

USP

Cyclohexylmethanol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cyclohexanemethanol, (Hydroxymethyl)cyclohexane, Cyclohexyl methanol, Hexahydrobenzyl alcohol, NSC 5288

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2OH
CAS Number:
Molecular Weight:
114.19
Beilstein:
773712
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benzyl alcohol

manufacturer/tradename

USP

refractive index

n20/D 1.465 (lit.)

bp

181 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCC1CCCCC1

InChI

1S/C7H14O/c8-6-7-4-2-1-3-5-7/h7-8H,1-6H2

InChI key

VSSAZBXXNIABDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyclohexylmethanol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Benzaldehyde

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service