G005907
HPLC Analysis of Aflatoxins in Wheat on Ascentis® Express C18 after SPE using Supel™ Tox AflaZea
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
sample preparation: SPE (Solid Phase Extraction)
sample/matrix: 25 g wheat meal spiked with pesticide standards
SPE tube/cartridge: Supel™Tox AflaZea, 6 mL (55314-U)
sample addition: 2 mL
elution: collect eluate in 5 mL silanized culture tubes
eluate post-treatment: combine 200 μL of sample eluate and 880 μL of DI water in a 2 mL autosampler vial
column: Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
mobile phase: [A] water; [B] methanol; [C] acetonitrile; (720:120:120, A:B:C) containing 0.780 g/L potassium bromide, 230 μL/L nitric acid
flow rate: 0.400 mL/min
pressure: 4568 psi (315 bar)
column temp.: 35 °C
detector: FLD 360 ex; 440 em
injection: 20 μL
derivatization: via KOBRA electrochemical cell, 50 μA
suitability
application for HPLC
application(s)
food and beverages
Analysis Note
Weigh 25 grams of wheat meal into a 500 mL Erlenmeyer flask. Add 100 mL of 84:16 acetonitrile:water, seal, and place on an orbital shaker at med-high speed for 90 minutes. Filter the sample through a #2 qualitative filter. Spike sample at 2 mg/mL using analytical standards.
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service