Skip to Content
Merck
All Photos(1)

Documents

89130

Supelco

(±)-Mevalonolactone-(methyl-d3)

analytical standard

Synonym(s):

Mevalonolactone-(methyl-d3), DL-Mevalonic-methyl-d3 lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D3H7O3
Molecular Weight:
133.16
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

bp

150 °C/5 mmHg (lit.)

mp

28 °C (lit.)

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C1(O)CCOC(=O)C1

InChI

1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3

InChI key

JYVXNLLUYHCIIH-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service