Skip to Content
Merck
All Photos(1)

Key Documents

808035

TSKgel® PWxl (10 µm) HPLC Columns

bottle of 1g, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Hardware and Accessories

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€138.00

€138.00


Please contact Customer Service for Availability


Select a Size

Change View
1 G
€138.00

About This Item

UNSPSC Code:
41115700

€138.00


Please contact Customer Service for Availability

Product Name

TSKgel® Top-Off for TSKgel® PWxl and G-DNA-PW columns, 10μm, 1g, phase hydroxylated methacrylate, 10 μm particle size, bottle of 1 g

Agency

suitable for USP L38

product line

TSKgel®

form

slurry

packaging

bottle of 1 g

manufacturer/tradename

Tosoh 08035

parameter

≤0.5 M salt concentration
≤20% organic solvent
1 mL/min flow rate
80 °C temperature

technique(s)

HPLC: suitable

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

10 μm

operating pH

2-12

separation technique

size exclusion (SEC)

General description

Top-Off for TSKgel PWxl and G-DNA-PW columns, 10 μm, 1 gram

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service