Skip to Content
Merck
All Photos(1)

Documents

13987-U

Supelco

Packed GC Column Set

suitable for 2177 per GPA (3-column set)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32110502
NACRES:
SB.54

Agency

suitable for GPA 2177 (3-column set)

technique(s)

gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

column type

packed GC

General description

Gas chromatography column are of basically two types packed column and open tubular column. In packed column the carrier gas must percolate through its porous bed. The shape and physical form of the packed column depends upon the dimensions of the chromatograph oven, the nature of the sample, the sample load and the column inlet pressure.
This column set is specifically designed for a specific column switching application. Column manufacturing procedurs have been optimized resulting in column sets that provide highly reproducible analyses.

Application

Packed GC column with flame ionization detector (FID) maybe used for the qualitative and quantitative analysis of gasoline and associated contaminated groundwater. It may also be used with electron capture detector (ECD) for the qualitative and quantitative analysis of halogenated compounds present in gasoline.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Introduction to Analytical Gas Chromatography.
Scott PW.
Science, 101-104 (1997)
Capillary and packed-column gas chromatography of gasoline hydrocarbons and EDB.
Bruell CJ and Hoag GE.
Science, 5-7 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service