Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2606

Sigma-Aldrich

Anti-HSPG2 Antibody, clone 1M17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Basement membrane-specific heparan sulfate proteoglycan core protein, HSPG, PLC, Perlecan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1M17
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 469 kDa
observed mol wt ~470 kDa

purified by

using Protein A

species reactivity

human, rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... HSPG2(3339)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1M17 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects HSPG2 (Perlecan). It targets an epitope within 25 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal region of human HSPG2 (Perlecan).

Application

Quality Control Testing

Evaluated by Western Blotting in Human umbilical vein endothelial cells (HUVEC) lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected HSPG2 in lysate from Human umbilical vein endothelial cells (HUVEC).

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected HSPG2 in lysates from SH-SY5Y and HeLa cells and Rat heart tissue.

Affinity Binding Assay: A representative lot of this antibody bound HSPG2 peptide with a KD of 1.1 x 10-9 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HSPG2 in HT-29 cells.

Immunohistochemistry (Paraffin) Analysis: 1:100 from a representative lot detected HSPG2 in Human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Basement membrane-specific heparan sulfate proteoglycan core protein (UniProt: P98160; also known as HSPG, Perlecan, PLC) is encoded by the HSPG2 gene (Gene ID: 3339) in human. HSPG2 is a large basement membrane heparan sulfate proteoglycan that is expressed in a wide array of tissues where it regulates diverse cellular processes including bone formation, inflammation, cardiovascular development, and angiogenesis. It is a major basement membrane component and colocalizes with elastin in blood vessels, which contributes to their visco-elastic properties in vasculogenesis during establishment of early vascular networks and tissue development. It is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to produce the mature protein that can be further proteolytically cleaved into Endorepellin (aa 3687-4391) and a LG3 peptide (aa 4197-4391). It is a modular multifunctional proteoglycan with five distinct domains and each domain has a specific function. Its protein core is divided into several unique structural regions, each imparting distinct biofunctional diversity. It is reported to interact with several other basement membrane components, including laminin, prolargin, and collagen type IV. It also displays a strong tendency to aggregate in dimers or stellate structures. Its N-terminal region contains the primary location for heparan sulfate side-chain attachment, and this region contains three areas comprising the signature Ser-Gly-Asp (SGD) sequence. Mutations in HSPG2 gene have been linked to Schwartz-Jampel syndrome, a rare autosomal recessive disorder that is characterized by permanent myotonia and skeletal dysplasia. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hayes, AJ., et al (2022). Front. Cell Dev. Biol. 10; 856261; Gubbiotti, MA., et al. (2017). Matrix Biol. 57-58: 285-298; Nicole, S., et al. (2000). Nat. Genet. 26(4); 480-483).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service