Skip to Content
Merck
All Photos(13)

Key Documents

ZRB2158

Sigma-Aldrich

Anti-IRF-5 Antibody, clone 1L11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Interferon regulatory factor 5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€191.00
4 X 25 μL
€393.00

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€191.00
4 X 25 μL
€393.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€191.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L11, recombinant monoclonal

description

1L11 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 56.04 kDa
observed mol wt ~56 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IRF5(3663)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone IL11 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Interferon regulatory factor 5 (IRF-5). It targets an epitope within 19 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal half of human Interferon regulatory factor 5 (IRF-5).

Application

Quality Control Testing

Evaluated by Western Blotting in LNCap cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IRF-5 in LNCap cell nuclear extract.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected IRF-5 in THP-1 and NIH3T3 cell nuclear extracts.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected IRF-5 in one million THP-1 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected IRF-5 in THP-1 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected IRF-5 in human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound IRF-5 peptide with a KD of 4.9 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in LNCap cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IRF-5 in LNCap cell nuclear extract.

Target description

Interferon regulatory factor 5 (UniProt: Q13568; also known as IRF-5) is encoded by the IRF5 gene (Gene ID: 3663) in human. The interferon regulatory factors (IRFs) are a family of nine transcription factors that regulate pathogen-induced innate and acquired immune responses. In unstimulated cells they reside in the cytoplasm. IRF-5 plays a critical role in innate immunity by activating expression of type I interferons (IFN- and β) and inflammatory cytokines downstream of endolysosomal toll-like receptors TLR7, TLR8, and TLR9. It regulates the transcription of IFN genes (IFNA and IFNB) and IFN-stimulated genes (ISG) by binding to an interferon-stimulated response element (ISRE) in their promoters. It is shown to be a key transcription factor regulating the interferon response during SARS-CoV-2 infection. It is maintained as a monomer in an autoinhibited state and undergoes homodimerization and activation following its phosphorylation on serine 451 and 462 in the C-terminal autoinhibitory region. It has been reported that phosphorylation of serine 451 relieves the autoinhibitory conformation, and phosphorylation of serine 462 stabilizes the IRF-5 homodimer. In its phosphorylation and activation scheme, the innate adapter protein TASL (TLR adaptor interacting with SLC15A4 on the lysosome) recruits IRF-5, which is then phosphorylated by IKK-β at serine 462 that allows IRF-5 dissociation from the adapter protein followed by its dimerization and nuclear translocation. Mutations in IRF5 gene have been linked to systemic lupus erythematosus 10 (SLEB10), rheumatoid arthritis, and irritable bowel syndrome. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Thompson, CD., et al. (2018). Front. Immunol. 9; 2622; Lopez-Pelaez, M., et al. (2014). Proc. Natl Acad. Sci. USA. 111(49); 17432-17437; Foreman, HCC., et al. (2012). PLoS One. 7(3); e33098).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service