Skip to Content
Merck
All Photos(1)

Key Documents

Z744021

Whirl-Pak® Hydrated PolyProbe

L 8 in., W × L 7.5 in. × 12 in.

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€401.00

€401.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€401.00

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

€401.00


Please contact Customer Service for Availability

packaging

pkg of 100 ea

manufacturer/tradename

B01592WA (NASCO)

L

8 in.

W × L

7.5 in. × 12 in.

General description

Designed for surface sampling and effective in hard-to-reach areas, the 8 in. inch (20.3 cm) long probe consists of a polypropylene holder with detachable handle that is adhered to a durable polyurethane sponge that resists tearing and fraying. Probe is packaged in a sealed 24-oz. (710 ml) write-on bag and hydrated with 10 ml of HiCap Neutralizing Broth. Product is terminally gamma processed to guarantee sterility. Box contains five foil pouches with 20 probes each for a total of 100 probes. Bags are 9 in. L x 6 in. W (23 x 15 cm) with a thickness of 2.5 mils (.064 mm). Two-year shelf-life with ambient storage at 36°-86° F (2°-30° C). To use, tear open the top of the bag, squeeze out any excess neutralizing broth, swab the surface, insert the sponge back into the bag, and, while holding the sponge from the outside, unscrew the handle and discard. Drop the sponge into the bag, fold the top of the bag over three or more times, bend in the wire tabs to close, and send to a lab for testing.

Legal Information

Whirl-pak is a registered trademark of The Aristotle Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service