Skip to Content
Merck
All Photos(1)

Key Documents

Z666548

Eppendorf® LoBind microcentrifuge tubes

DNA/RNA, volume 1.5 mL

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
eCl@ss:
32119210

material

cap (attached)
polypropylene
transparent tube

sterility

non-sterile

feature

RCF 18,000 × g
RNase and DNase free

packaging

pack of 250 (5 bags of 50)

manufacturer/tradename

Eppendorf® 0030 108.051 (European)
Eppendorf® 022431021 (USA)

volume

1.5 mL

Looking for similar products? Visit Product Comparison Guide

General description


  • Minimized protein loss
  • Free of surface coating, e.g., silicone
  • Free of DNA, DNase, RNase and ATP (PCR clean)
  • Special polypropylene material manufactured under optimized processing conditions
  • Available in 0.5 mL, 1.5 mL and 2.0 mL sizes
  • Rated to 18,000 x g
  • High degree of transparency
Protein LoBind tubes are specifically designed for use in proteomics or other fields of protein research (antibodies, cell biology, peptide drugs etc.), where samples tend to be very small and sample recovery is vital. LoBind tubes improve recovery by reducing sample-to-tube binding—without the use of any additives or coatings! A combination of specially developed technology and the purest, select polypropylene results in tube binding that ensures minimal loss (<3%). Enzymes remain active, and they are not denatured by any tube-surface interactions.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid M Alsema et al.
Frontiers in molecular neuroscience, 13, 134-134 (2020-11-17)
Microglia are the tissue-resident macrophages of the central nervous system (CNS). Recent studies based on bulk and single-cell RNA sequencing in mice indicate high relevance of microglia with respect to risk genes and neuro-inflammation in Alzheimer's disease (AD). Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service