Key Documents
WPB01041A
Whirl-Pak® Sampling Spoons
sterile
Synonym(s):
dispensing tool
Select a Size
WPB01041A-100EA
€28.20
1000 EA
€249.00
€28.20
Please contact Customer Service for Availability
Select a Size
Change View
WPB01041A-100EA
€28.20
1000 EA
€249.00
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
€28.20
Please contact Customer Service for Availability
material
polystyrene
sterility
sterile
packaging
pkg of 100 ea
manufacturer/tradename
Nasco Whirl-pak® B01041A
L
~18 cm
capacity
~15 mL
General description
This sterilized spoon is ideal for use with Whirl-Pak® bags where sterile techniques are required for collection of samples.
Features and Benefits
  • Single-time use removes contamination risk
  • The easy-to-use spoon is made of sturdy, durable polystyrene
  • The procedure for contamination-free use is to open the bag, push one spoon up, pull it out, and reclose the bag
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)