Skip to Content
Merck
All Photos(6)

Key Documents

WH0002936M1

Sigma-Aldrich

Monoclonal Anti-GSR antibody produced in mouse

clone 6B4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MGC78522, Anti-glutathione reductase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6B4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GSR(2936)

Immunogen

GSR (NP_000628, 413 a.a. ~ 522 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TVVFSHPPIGTVGLTEDEAIHKYGIENVKTYSTSFTPMYHAVTKRKTKCVMKMVCANKEEKVVGIHMQGLGCDEMLQGFAVAVKMGATKADFDNTVAIHPTSSEELVTLR

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Wu et al.
Clinical and experimental immunology, 176(3), 438-451 (2014-02-18)
Innate immune and inflammatory responses are involved in myocardial ischaemia/reperfusion (I/R) injury. Interleukin (IL)-37 is a newly identified member of the IL-1 family, and functions as a fundamental inhibitor of innate immunity and inflammation. However, its role in myocardial I/R
Mohammed Merza et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(9), G914-G921 (2014-09-13)
Severe acute pancreatitis (AP) is characterized by leukocyte infiltration and tissue injury. Herein, we wanted to examine the potential effects of thrombin-derived host defense peptides (TDPs) in severe AP. Pancreatitis was provoked by infusion of taurocholate into the pancreatic duct
Marc-André Wurbel et al.
Inflammatory bowel diseases, 20(7), 1165-1176 (2014-05-31)
The chemokine CCL25, and its receptor CCR9, constitute a unique chemokine/receptor pair, which regulates trafficking of T lymphocytes to the small intestine under physiological conditions and is an attractive target for small bowel Crohn's disease drug development. We have previously
Hitoshi Ishiguro et al.
American journal of clinical pathology, 142(2), 157-164 (2014-07-13)
To assess the levels of glucocorticoid receptor (GR) expression in bladder tumors because the status and its prognostic value remain largely unknown. We immunohistochemically stained for GR in bladder tumor and matched non-neoplastic bladder tissue specimens. Overall, GR was positive
Milladur Rahman et al.
Platelets, 25(4), 257-263 (2013-07-17)
Abstract Platelets play an important role in abdominal sepsis and P2Y12 receptor antagonists have been reported to exert anti-inflammatory effects. Herein, we assessed the impact of platelet inhibition with the P2Y12 receptor antagonist ticagrelor on pulmonary neutrophil recruitment and tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service