Skip to Content
Merck
All Photos(1)

Documents

VIV102012

SynVivo SynTumour 3D cancer model chip, IMN2 radial configuration, 8 um pillar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NA.84

packaging

case of 1 ea

manufacturer/tradename

SynVivo 102012

storage temp.

room temp

General description

SynTumor cancer model chip-Barrier type: Pillars, Distance between pillars: 8 μm, Barrier height: 8 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 um. SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment.

Linkage

LInk to Publications
application text

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service