Key Documents
TMO3664-0100
Nalgene® economy graduated cylinder
polypropylene, volume 100 mL, case of 12 ea
Synonym(s):
PP graduated cylinder, PP graduated flask
Select a Size
12 EACH
€123.00
€123.00
Please contact Customer Service for Availability
Select a Size
Change View
12 EACH
€123.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€123.00
Please contact Customer Service for Availability
material
polypropylene
packaging
case of 12 ea
manufacturer/tradename
Nalgene 3664-0100
capacity
100 mL
volume
100 mL
General description
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)