Skip to Content
Merck
All Photos(1)

Documents

TMO312009-0008

Nalgene® rectangular bottle

high-density polyethylene, bottle volume 250 mL, amber, case of 250 ea

Synonym(s):

light-sensitive storage bottle, opaque Amber bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

high-density polyethylene
rectangular bottle

packaging

case of 250 ea

manufacturer/tradename

Nalgene 312009-0008

bottle volume

250 mL

color

amber

Looking for similar products? Visit Product Comparison Guide

General description

Protect light-sensitive liquids or dry materials from exposure with Thermo Scientific Nalgene Rectangular Opaque Amber HDPE Bottles with Closure. Protection from UV light combines with a space-saving shape. Uniform high density polyethylene walls are resistant to splitting or puncturing and have a excellent chemical resistance to most acids, bases and alcohols. Bottles offer a leakproof† sealing when used with opaque polypropylene Nalgene closures.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service