Skip to Content
Merck
All Photos(1)

Key Documents

SRP3099

Sigma-Aldrich

IL-9 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

P40 cytokine, T-cell growth factor p40

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€329.00

€329.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€329.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€329.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤1 ng/mL ED50

mol wt

14.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL9(3578)

General description

IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. IL-9 enhances the proliferation of T lymphocytes, mast cells, erthroid precursor cells and megakaryoblastic leukemia cell lines. Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin′s disease. Whereas murine IL-9 can function on human cells, human IL-9 is inactive on mouse cells. Recombinant human IL-9 is a 14.0 kDa protein of 127 amino acid residues including 10 cysteine residues which are fully conserved between the human murine proteins.

Biochem/physiol Actions

IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. Recombinant human IL-9 is a 14.0 kDa protein of 127 amino acid residues including 10 cysteine residues which are fully conserved between the human murine proteins.

Sequence

MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI

Physical form

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fitzgerald, K.A., et al.
The Cytokine Factsbook, 85-89 (2001)
K Kelleher et al.
Blood, 77(7), 1436-1441 (1991-04-01)
We have isolated the genomic sequence of human interleukin-9 (IL-9) based on its sequence homology with a human IL-9 cDNA isolated from human T-cell leukemia virus (HTLV)-I-transformed T cells by expression cloning. The entire genomic sequence has been determined and
Gabriella Ruocco et al.
Clinical science (London, England : 1979), 129(4), 291-303 (2015-02-24)
Multiple sclerosis (MS) is a chronic disease of the central nervous system (CNS) characterized by persistent inflammation orchestrated by cluster of differentiation (CD) 4 T helper (Th) cells. In particular, Th1 and Th17 cells amplify, whereas T regulatory (Treg) cells
Juan Liu et al.
The Journal of investigative dermatology, 134(7), 1903-1911 (2014-02-04)
The cytokine IL-9, derived primarily from T-helper 9 (Th9) lymphocytes, promotes expansion of the Th2 subset and is implicated in the mechanisms of allergic asthma. We hypothesize that IL-9 also has a role in human allergic contact dermatitis (ACD). To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service