Skip to Content
Merck
All Photos(1)

Key Documents

SRP2159

Sigma-Aldrich

Rev-ErbA, β (RVR) human

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Synonym(s):

BD73, EAR-1R, RVR

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€470.00

€470.00


Please contact Customer Service for Availability


Select a Size

Change View
5 μG
€470.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€470.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in insect cells

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~66.6 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

350 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NR1D2(9975)

Biochem/physiol Actions

Nuclear receptors form the largest known family of transcription factors and have a crucial role in nearly all aspects of vertebrate development and adult physiology by transducing the effects of hormones into transcriptional responses. Members of the steroid/thyroid hormone nuclear receptor (NR) superfamily bind specific DNA elements and function as ligand activated transcription factors. This group includes the `orphan receptors′ which have no known ligands in the `classical sense′ and appear to be the ancient progenitors of this receptor superfamily. The Rev-erb family of proteins are orphan members of the receptor superfamily. Two isoforms of the Rev-erb family have been isolated from mammalian genotypes, Rev-erbA[alpha] and Rev-erbA[beta]/RVR. Major differences between the two isoforms occur within the hyper-variable A/B and D regions of the proteins. Both isoforms are expressed in a wide range of tissues and are present in all major organs. Rev-erbA[alpha] mRNA is upregulated during adipocyte differentiation but repressed during myogenesis. These orphan receptors are closely related to the ROR/RZR[alpha] gene family (retinoic acid receptor related orphan receptor) and the Drosophila orphan receptor, E75A, particularly in the DNA-binding domain (DBD) and the putative ligand-binding domain (LBD). RVR and Rev-erbA[alpha] bind as monomers to an asymmetric ( A / T ) 6 RGGTCA motif. The Rev-erb family has also been demonstrated to bind as homodimers to novel HREs consisting of two tandemly arranged AGGTCA motifs, separated by 2 bp with unique 5′ flanking and spacer nucleotides (RevDR-2). Reports on the transcriptional properties of the Rev-erb family were initially conflicting. Rev-erbA[alpha] was first reported to act as a constitutive activator of transcription through its cognate monomeric asymmetric motif. Recently, it has been demonstrated that members of the Rev-erb family are, in fact, dominant repressors of transcription. Rev-erbA[beta] is expressed in the central nervous system, skeletal and dorsal muscles, spleen and mandibular and maxillar processes. During embryogenesis RVR is expressed in the notochord and neural tube, but its function/role during differentiation and mammalian development remains obscure.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Evans
Science (New York, N.Y.), 240(4854), 889-895 (1988-05-13)
Analyses of steroid receptors are important for understanding molecular details of transcriptional control, as well as providing insight as to how an individual transacting factor contributes to cell identity and function. These studies have led to the identification of a
M A Lazar et al.
Molecular and cellular biology, 9(3), 1128-1136 (1989-03-01)
A cDNA encoding a novel member of the thyroid/steroid hormone receptor superfamily, called Rev-ErbA alpha, has been isolated from a rat GH3 cell library. Rev-ErbA alpha is an approximately 56-kilodalton protein most similar in structure to the thyroid hormone receptor
The nuclear receptor superfamily: the second decade.
D J Mangelsdorf et al.
Cell, 83(6), 835-839 (1995-12-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service