Skip to Content
Merck
All Photos(1)

Key Documents

SML3545

Sigma-Aldrich

NVP-RXI570

≥98% (HPLC)

Synonym(s):

1-[3-(Difluoromethyl)isothiazol-5-yl]-3-[2-fluoro-5-(hydroxymethyl)pyridin-4-yl]urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€95.60
25 MG
€385.00

€95.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€95.60
25 MG
€385.00

About This Item

Empirical Formula (Hill Notation):
C11H9F3N4O2S
CAS Number:
Molecular Weight:
318.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

€95.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(F)=NC=C1CO)NC2=CC(C(F)F)=NS2

InChI

1S/C11H9F3N4O2S/c12-8-1-6(5(4-19)3-15-8)16-11(20)17-9-2-7(10(13)14)18-21-9/h1-3,10,19H,4H2,(H2,15,16,17,20)

InChI key

CKYCAIAVJIFWPE-UHFFFAOYSA-N

Biochem/physiol Actions

NVP-RXI570 is an orally active, potent, and selective allosteric dual inhibitor of BRM ((SMARCA2) and BRG1 (SMARCA4) that that downregulates BRM-dependent gene expression. NVP-RXI570 exhibits antiproliferative activity in a BRG1 mutant-lung-tumor xenograft model.
Orally active, potent, and selective allosteric dual inhibitor of BRM ((SMARCA2) and BRG1 (SMARCA4)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of Orally Active Inhibitors of Brahma Homolog (BRM)/SMARCA2 ATPase Activity for the Treatment of Brahma Related Gene 1 (BRG1)/SMARCA4-Mutant Cancers
Journal of medicinal chemistry, 61, 10155-10172 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service