Skip to Content
Merck
All Photos(1)

Documents

SML3466

Sigma-Aldrich

JHU37160

≥98% (HPLC)

Synonym(s):

8-Chloro-11-(4-ethylpiperazin-1-yl)-4-fluoro-5H-dibenzo[b,e][1,4]diazepine, J60, JH60, JHU 37160, JHU-37160

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClFN4
CAS Number:
Molecular Weight:
358.84
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

JHU37160 (J60) is a potent DREADDs (Designer Receptors Exclusively Activated by Designer Drugs) ligand (hM3Dq/hM4Di Ki = 1.9/3.6 nM using mouse brain tissue, 18.5/0.2 nM by celluar BRET assays) with ~25-fold improved affinity than C21 and high in vivo potency for CNS applications. J60 selectively competes against clozapine at DREADDs, but not other clozapine-binding sites in mouse brain tissue ex vivo (10 nM), increases hM3Dq-stimulated locomotion in TH-hM3Dq rats (0.01-0.3 mg/kg i.p.) and selectively inhibits locomotor activity of D1-hM3Dq and D1-hM4Di transgenic, but not wild-type, mice (0.01-1 mg/kg i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service