Skip to Content
Merck
All Photos(1)

Documents

SML0728

Sigma-Aldrich

ANR-94

≥98% (HPLC)

Synonym(s):

8-Ethoxy-9-ethyl-9H-purin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5O
CAS Number:
Molecular Weight:
207.23
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

room temp

InChI

1S/C9H13N5O/c1-3-14-8-6(7(10)11-5-12-8)13-9(14)15-4-2/h5H,3-4H2,1-2H3,(H2,10,11,12)

InChI key

QUGDTMONBLMLLD-UHFFFAOYSA-N

Biochem/physiol Actions

ANR-94 is a potent antagonist of the adenosine A2A receptor (AA2AR). The Ki for human AA2AR is 46 nM. ANR-94 displays neuroprotective properties in rat models of Parkinson′s disease. ANR-94 reverses GABA-A receptor desensitization in oocyte and neuronal electrophysiological preparations.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to adenosine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service