Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702771

Sigma-Aldrich

Anti-CDK8 Antibody, clone 7L5X2, Rabbit Monoclonal

Synonym(s):

K35

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€378.00

€378.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€378.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€378.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

7L5X2, monoclonal

form

liquid

mol wt

55 kDa

species reactivity

human, rat, rat

concentration

0.8 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MDYDFKVKLSSERERVEDLFEYEGCKVGRGTYGHVYKAKRKDGKDDKDYALKQIEGTGISMSACREIALLRELKHPNVISLQKVFLSHADRKVWLLFDYA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK8(1024)

General description

This gene encodes a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are known to be important regulators of cell cycle progression. This kinase and its regulatory subunit, cyclin C, are components of the Mediator transcriptional regulatory complex, involved in both transcriptional activation and repression by phosphorylation of the carboxy-terminal domain of the largest subunit of RNA polymerase II. This kinase regulates transcription by targeting the cyclin-dependent kinase 7 subunits of the general transcription initiation factor IIH, thus providing a link between the Mediator complex and the basal transcription machinery. Multiple pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2016]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDK8 (P49336).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service