Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702561

Sigma-Aldrich

Anti-BHMT Antibody, clone 7O7D7, Rabbit Monoclonal

Synonym(s):

BHMT1, HEL-S-61p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

7O7D7, monoclonal

form

liquid

mol wt

45 kDa

species reactivity

rat, rat, human

concentration

0.56 mg/mL

technique(s)

immunohistochemistry: 1:500 - 1:700
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

CGFEPYHIRAIAEELAPERGFLPPASEKHGSWGSGLDMHTKPWVRARARKEYWENLRIASGRPYNPSMSKPDGWGVTKGTAELMQQKEATTEQQLKELFEKQKFKSQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BHMT(635)

General description

This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 300-406 of human BHMT (Q93088).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service