Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702050

Monoclonal Anti-CD14 APC-Cyanine7 produced in mouse

new

Synonym(s):

LPS-receptor, LPSR, MCLR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

APC-Cy7 conjugate

Quality Level

antibody product type

primary antibodies

clone

MEM-15, monoclonal

form

buffered aqueous solution

species reactivity

( Human), (Non-human primates)

technique(s)

flow cytometry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD14(929)

General description

CD14 is a 55 kDa GPI-anchored glycoprotein, constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils, where it serves as a multifunctional lipopolysaccharide receptor. It is also released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 is able to discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. CD14 affects allergic, inflammatory and infectious processes.

Specificity

The antibody MEM-15 reacts with CD14, a 53-55 kDa GPI (glycosylphosphatidylinositol)-linked extracellular membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. The antibody MEM-15 also reacts with soluble forms of CD14 found in serum and in the urine of some nephrotic patients.

Immunogen

A crude mixture of human urinary proteins precipitated by ammonium sulphate from the urine of a patient suffering from proteinuria.

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with activated tandem dye of allophycocyanin-cyanine 7 (APC-Cy7) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service